Would you like to log in again?
CARTRIDGE FLASHPURE SELECT C18 12G BX5
Supplier: Buchi
FlashPure Select C18 catridge 12g^ FlashPure Select C18 catridge, Spherical, Particle size: 20-35um, Pore size: 92-108A, Max.Pressure: 180psi / 12bar, Carbon content: 17-19%, with End capping, Default flowrate: 30 mL/min, Loading capacity: 12mg-0.6g, Column volume: 24 mL, weight: 12g
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CARTRIDGE FLASHPURE SELECT C18 12G BX5 76196-942 140000122 Box of 5 (12g) Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty