FlashPure cartridges are offered in a wide range of sizes, covering different stationary phases, particle sizes and geometries. This enables the user to choose the flash cartridge which best suits his purification needs.
- Different sizes for scalability from milligrams to grams
- Broad range of different normal and reversed phases
- Small and spherical particles for fast separations and high loadings
Compatibility with the Pure Navigator software (flash method optimization by interpreting TLC or HPLC scouting runs). RFID tag for an automatic transfer of important parameters for a run on a Pure chromatography system.
Silica phase are high and medium polar compounds, most popular polar phase, mainly used for flash applications. C18 WP (wide pores) has a ideal application in peptides.
C18 phase are low polar and non-polar compounds, most popular non-polar phase. Amino phase are high and medium polar molecules example carbohydrates and amines. Alumina Wide variety of molecules example acidic, basic or acid sensitive compounds.
Diol, C18 and Amino phase has spherical particle shape. Silica and alumina has irregular shape.
Loading capacity: 0.1 to 10% for Silica, Silica LOK, Silica Solid Loader; 0.1 to 4% for Alumina Neutra, Alumina Basic, Alumina Acidic; 0.1 to 2.5% for C18; 0.1 to 5% for Amino, Diol.