Would you like to log in again?
KIMTECH PURE® A8 Certified Lab Coat + Extra Protection, Kimberly Clark Professional
Supplier: Kimberly-Clark
10049
76199-146CS 269.48 USD InStock
76199-146
KIMTECH PURE® A8 Certified Lab Coat + Extra Protection, Kimberly Clark Professional
Disposable Garments
Disposable, fluid-resistant lab coats are manufactured from a protective three-layer spunbond-meltblown-spunbond (SMS) fabric that offers exceptional resistance to blood strikethrough for moderate fluid contact.
  • Breathable, three-layer SMS fabric
  • Extra Protection offers blood strikethrough resistance
  • Soft knit cuffs for added comfort and protection
  • Nonsterile
  • Latex-free
Lab coats feature a six-snap front closure, three reinforced pockets, back vent, traditional collar, and knit cuffs. Tailored styling and fit enhances worker comfort.
Recyclable through the Rightcycle Program.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty