Would you like to log in again?
Workbrutes® PVC Overshoes, Tingley Rubber
Supplier: Tingley
Workbrutes®
35111-2X 35111-XL 35111-XS 35111-M 35111-SM 35111-L
76199-748CS 148.03 USD InStock
76199-748 76199-154 76199-156 76199-158 76199-744 76199-746
Workbrutes® PVC Overshoes, Tingley Rubber
Shoes and Boots
PVC Overshoes with thicker, deeper cleats for better grip and longer wear.
  • Covers work shoe up to ankle
  • Heavy duty heel kick off for easy off
Frigiflex® PVC compound contains special additives for high stretch and flexibility in cold temperatures. The seamless construction provides 100% waterproof protection.
Cleated outsole spits out debris and provides excellent traction. The injection molding puts less material in the upper, eliminating unnecessary weight, for reduced leg fatigue. Engineered for a longer life by adding more material to the sole at the greatest wear points: the heel, toe and cleats. Key interior entry points are specially textured to allow overshoes to easily slip on and off over rubber sole shoes.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty