Would you like to log in again?
PLATE PCR 96WELL HALF SKIRT PP CLR CS400
Supplier: Greiner Bio-One
PCR Plate 96 well^ PCR Plate, well density: 96well, color: Clear, Format: half skirt, Rigid Style, without Lid, 96well & 384 well formats, for ultra high throughput DNA analysis, Highly visible alphanumerics, Compatible with variety of instruments, including ABI and LightCycler
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
PLATE PCR 96WELL HALF SKIRT PP CLR CS400 76199-204 652235 Case of 400 Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty