Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 2PM ET on 20th April 2024.
VWR® ESD Micro Denier Scrub Top with Knit Cuff
Supplier: VWR
VWR®
2039ESW1NVY1XL 2039ESW1NVY3XL 2039ESW1NVY2XL 2039ESW1NVYXXS 2039ESW1NVY5XL 2039ESW1NVY6XL 2039ESW1NVY4XL 2039ESW1NVYS 2039ESW1NVYM 2039ESW1NVYL 2039ESW1NVYXS
76202-672EA 54.84 USD InStock
76202-672 76202-660 76202-674 76202-662 76202-676 76202-664 76202-678 76202-666 76202-658 76202-668 76202-670
VWR® ESD Micro Denier Scrub Top with Knit Cuff
Workwear
This style of scrub top is widely used in pharmaceutical cleanroom operations, and meets/exceeds the recommendations outlined in the IEST-RP-CC-003.4, Garment Considerations for Cleanrooms and Other Controlled Environments.
  • Pullover design with three-snap, Henley-style collar that provides added room for easy on/off
  • Binding on collar helps maintain shape and uniformity
  • Raglan-style sleeves provide increased mobility
  • Sleeves include color-coordinated filament knit cuff for extra comfort
  • 5" side slits at garment bottom for added comfort and long-term durability
These scrubs provide an excellent base-layer of protection for any cleanroom garment system. Made with Sandwash material, this lightweight micro-denier fabric is woven from 100% continuous-filament polyester to eliminate any source for lint.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty