Would you like to log in again?
Human Emmprin ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0443
76235-604EA 687 USD InStock
76235-604
Human Emmprin ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human Emmprin AccuSignal ELISA Kit
  • Natural and recombinant human Emmprin.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: T25-H205
Basigin is a member of the immunoglobulin superfamily that is also known as EMMPRIN, short for extracellular matrix metalloproteinase inducer, and recently has been designated CD147(cluster of differentiation 147). It is a member of the immunoglobulin superfamily, with a structure related to the putative primordial form of the family. As members of the immunoglobulin superfamily play fundamental roles in intercellular recognition involved in various immunologic phenomena, differentiation, and development, basigin is thought also to play a role in intercellular recognition(Miyauchi et al., 1991; Kanekura et al., 1991).This protein is a determinant for the Ok blood group system. Basigin is a type I integral membrane receptor that has many ligands, including the cyclophilin(CyP) proteins Cyp-A and CyP-B and certain integrins. It is expressed by many cell types, including epithelial cells, endothelial cells and leukocytes.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml human Emmprin standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum, plasma( heparin, EDTA), saliva, urine or human milk to each empty well. It is recommended that each human Emmprin standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty