Would you like to log in again?
Mouse Clusterin ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0559
76235-682EA 687 USD InStock
76235-682
Mouse Clusterin ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse Clusterin AccuSignal ELISA Kit
  • Natural and recombinant mouse Clusterin.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: D22-R226(beta)&S227-E448(alpha)
Clusterin(apolipoprotein J) is a 75 - 80 kDa disulfide-linked heterodimeric protein associated with the clearance of cellular debris and apoptosis.[1] In humans, clusterin is encoded by the CLU gene.This protein has several synonyms: dimeric acidic glycoprotein(DAG protein), testosterone repressed prostate message-2(TRPM-2), sulfated glycoprotein-2(SGP-2) and complement lysis inhibitor(CLI). Clusterin was mapped to mouse chromosome 14. Clusterin is a ubiquitously expressed molecule thought to influence a variety of processes including cell death. In the brain, it accumulates in dying neurons following seizures and hypoxic-ischemic(H-I) injury. clusterin may be a new therapeutic target to modulate non-caspase-dependent neuronal death following acute brain injury.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 20000pg/ml, 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml mouse Clusterin standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse Clusterin standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty