Would you like to log in again?
Human ANGPTL4 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0590
76235-718EA 636 USD InStock
76235-718
Human ANGPTL4 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human ANGPTL4 AccuSignal ELISA Kit
  • Natural and recombinant human ANGPTL4.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: L165-S406
Angiopoietin-related protein 4(Angptl4) is a protein that in humans is encoded by the ANGPTL4 gene. This gene is a member of the angiopoietin/angiopoietin-like gene family and encodes a glycosylated, secreted protein with a fibrinogen C-terminal domain. This gene is induced under hypoxic conditions in endothelial cells and is the target of peroxisome proliferation activators. By radiation hybrid analysis, Angptl4 gene is mapped to 19p13.3. ANGPTL4 contributed to tumor growth and protected cells from anoikis, a form of programmed cell death induced when contact-dependent cells detach from the surrounding tissue matrix.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 40000pg/ml, 20000pg/ml, 10000pg/ml, 5000pg/ml, 2,500pg/ml, 1,250pg/ml, 625pg/ml human ANGPTL4 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human ANGPTL4 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty