Would you like to log in again?
Human FASL ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0165
76236-598EA 713 USD InStock
76236-598
Human FASL ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human FASL AccuSignal ELISA Kit
  • Natural and recombinant human FASL.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: CHO; Immunogen sequence: P134-L281
Fas ligand(FasL or CD95L) is a type-II transmembrane protein that belongs to the tumor necrosis factor(TNF) family. Its binding with its receptor induces apoptosis. The human FASL gene consists of approximately 8 kb and is split into 4 exons. Fas ligand/receptor interactions play an important role in the regulation of the immune system and the progression of cancer. Fas ligand or FasL is a homotrimeric type II transmembrane protein. It signals through trimerization of FasR, which spans the membrane of the "target" cell. This trimerization usually leads to apoptosis, or cell death. Soluble Fas ligand is generated by cleaving membrane-bound FasL at a conserved cleavage site by the external matrix metalloproteinase MMP-7.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml human FASL standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA, citrate) to each empty well. It is recommended that each human FASL standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty