Would you like to log in again?
Human CCL21 - 6Ckine ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0335
76236-832EA 636 USD InStock
76236-832
Human CCL21 - 6Ckine ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human CCL21 - 6Ckine AccuSignal ELISA Kit
  • Natural and recombinant human CCL21.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: S24-P134
Chemokine(C-C motif) ligand 21(CCL21) is a small cytokine belonging to the CC chemokine family. This chemokine is also known as 6Ckine(because it has six conserved cysteine residues instead of the four cysteines typical to chemokines), exodus-2, and secondary lymphoid-tissue chemokine(SLC). By somatic cell hybrid and radiation hybrid analyses, mapped the SCYA21 gene to 9p13. Chemokines are a family of proteins that direct leukocyte migration and activation to inflammatory stimuli. CXC chemokine ligand 13(CXCL13), CC chemokine ligand 21(CCL21), and CCL19 are constitutively expressed in secondary lymphoid organs, where they control the placement of lymphocytes and dendritic cells. It was demonstrate that the local expression of homeostatic chemokines in nonlymphoid organs, such as the lung, plays an important role in protective immune responses.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 4000pg/ml, 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml human CCL21 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human CCL21 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty