Would you like to log in again?
Human Ficolin-1 ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0741
76236-974EA 840 USD InStock
76236-974
Human Ficolin-1 ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Human Ficolin-1 AccuSignal ELISA Kit
  • Natural and recombinant human Ficolin-1.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: A30-A326
Ficolin-1, and also commonly termed M-ficolin is a protein that in humans is encoded by the FCN1 gene. It is mapped to 9q34.3. The ficolin family of proteins are characterized by the presence of a leader peptide, a short N-terminal segment, followed by a collagen-like region, and a C-terminal fibrinogen-like domain. Ficolins have a crucial role in defense against pneumococcal infection through the lectin complement pathway. Ficolin 1 is predominantly expressed in the peripheral blood leukocytes, and has been postulated to function as a plasma protein with elastin-binding activity.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 50ng/ml, 25ng/ml, 12.5ng/ml, 6.25ng/ml, 3.12ng/ml, 1.56ng/ml, 0.78ng/ml human Ficolin-1 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin) to each empty well. It is recommended that each human Ficolin-1 standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty