Would you like to log in again?
Mouse IL-17D ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
KOA0467
76237-320EA 636 USD InStock
76237-320
Mouse IL-17D ELISA Kit, Rockland Immunochemicals
Assays ELISAs
Mouse Interleukin-17D AccuSignal ELISA Kit
  • Natural and recombinant mouse IL-17D.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.colI; Immunogen sequence: A25-R205
Interleukin 17D (IL-17D) is a member of the unique IL-17 family, which share a conserved cysteine motif. It is mapped to 13q12.11. IL-17D regulates cytokine production in endothelial cells and inhibits hemopoiesis in vitro. Purified recombinant IL-17D inhibited myeloid progenitor cell colony formation. IL-17D can enhance endothelial cell production of IL6, IL8, and GMCSF, but not other cytokines. And it is preferentially expressed in skeletal muscle, brain, adipose tissue, heart, lung, and pancreas.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 4000pg/ml, 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml mouse IL-17D standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse IL-17D standard solution and each sample be measured in duplicate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty