Would you like to log in again?
ESD-Safe Mat Cleaner, Jensen Tools
Supplier: Jenson Tools
8001
76238-096EA 40.69 USD InStock
76238-096
ESD-Safe Mat Cleaner, Jensen Tools
Cleaning Agents Detergents and Surface Cleansers
Extra strength cleaner for removing dirt, grime and stains from all types of static control surfaces, including table and floor mats, hard laminates, bench tops and portable field service kits.
  • Surface mark remover
  • Fresh, clean scent
  • Water-based liquid
  • Leaves no tacky or corrosive residue
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty