Would you like to log in again?
Saturix C30 Polyester/Cellulose Wipes
Supplier: FG Cleanwipes
Saturix
7C30BBP04 7C30HHP00 7C3099P09
76240-468CS 1175.93 USD InStock
76240-468 76238-846 76238-842
Saturix C30 Polyester/Cellulose Wipes
Wipes Cleanroom Wipes Dry Cleanroom Wipes
These polyester/cellulose wipes offer the essential benefits of a non-woven wipe, high absorbency with aqueous liquids, dependable durability, and value. The exacting manufacturing process combines versatility, economy, and cleanliness.
  • Hydroentangled blend of 55% cellulose and 45% polyester fabric (64 g/m²)
  • Super absorbent
  • Extremely low particle generation
  • No chemical binders
Economical, general-purpose wipes offer extremely low particle generation with no chemical binders. Manufactured with a standard weight, hydroentangled blend of 55% cellulose and 45% polyester, C30 wipes are highly absorbent and residue-free, with low NVRs and ion extractalbes. Recommended for use in ISO 5 to 8 controlled environments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty