Would you like to log in again?
Lime Remover Cleaners, Sunburst Chemicals
Supplier: Sunburst
66224
76253-934CS 313.8 USD InStock
76253-934
Lime Remover Cleaners, Sunburst Chemicals
Cleaning Agents Detergents and Surface Cleansers
Effective on stainless steel, ceramic, glass and plastic: contains no muriatic, hydrochloric or sulfuric acid.
  • Non-foaming
  • No sharp odors or fumes
Super concentrated ingredients and penetrating agents provide maximum effectiveness.
Continued regular use helps your dishwasher, steam table, coffee maker and ice machine perform at maximum efficiency.
Breaks down hard water scale and removes rust stains from fixtures or equipment.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty