Would you like to log in again?
RACK W/CAPPED TUBES PC BLUE 300#L CS960
Supplier: Greiner Bio-One
Rack W/Capped Tubes Pc Blue 300ul Cs960^ Rack, with 96 Cryo.s* Biobanking capped Tubes, with Screw cap, Material: Polypropylene(tube/cap), Polycarbonate(rack/lid), Gasket: Silicone, medical grade, Color: blue(cap), natural(tube), 5 racks/bag, 1 manual cap
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
RACK W/CAPPED TUBES PC BLUE 300#L CS960 76292-228 976584 Case of 960 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty