Would you like to log in again?
Vesphene® III se Phenolic Disinfectants
Supplier: Steris
Vesphene® III
647508 647501 647505
76303-114EA 484.43 USD InStock
76303-114 76303-152 76303-116
Vesphene® III se Phenolic Disinfectants
Cleaning Agents Disinfectants
STERIS' Vesphene® III se Phenolic disinfectant is a liquid concentrate specifically formulated for the disinfection of hard surfaces. The use-dilution can be applied manually or sprayed.
  • Efficacy - Phenolic active ingredients provide broad spectrum antimicrobial efficacy
  • Productivity - Formulation includes surfactants for surface cleaning while disinfecting
  • Flexibility - Ideal for use in a contamination control rotation
  • Reliability - Complete lot traceability
  • Assurance - Each lot documented by Certificate of Analysis
STERIS' Vesphene® III se disinfectant is specifically formulated for the routine disinfection of hard, non-porous surfaces. At a use dilution of rate of 0.8% v/v (1:128), the Vesphene® III disinfectants kill a broad spectrum of common bacteria and fungi. The alkaline Vesphene® III disinfectant formulation can be used alone or in rotation with the LpH®® III disinfectants. These products have an extensive documentation package to meet your validation objectives and are supported by the STERIS Technical Services team for individualized application and validation assistance.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty