Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GRK2 Goat Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-101-MH5 600-101-MH5S
76309-342EA 148 USD InStock
76309-342 76309-340
Anti-GRK2 Goat Polyclonal Antibody
Antibodies
GRK2 (G Protein-Coupled Receptor Kinase 2) is a member of the G protein-coupled receptor kinase family of proteins. GRK2 Specifically phosphorylates the agonist-occupied form of the beta-adrenergic and as well as a wide range of other substrates including non-GPCR cell surface receptors, and cytoskeletal, mitochondrial, and transcription factor proteins. It is a key regulator of LPAR1 signaling. GRK2 competes with RALA for binding to LPAR1 thus affecting the signaling properties of the receptor. It desensitizes LPAR1 and LPAR2 in a phosphorylation-independent manner. And positively regulates ciliary smoothened (SMO)-dependent Hedgehog (Hh) signaling pathway by facilitating the trafficking of SMO into the cilium and the stimulation of SMO activity. Data from rodent models supports a role for this gene in embryonic development, heart function and metabolism. Elevated expression of this gene has been observed in human patients with heart failure and Alzheimer's disease. Anti-GRK2 Antibody is useful for researchers interested in Neuroscience, Cytokines & Growth Factors, and Stem Cell Research.

Type: Primary
Antigen: GRK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty