Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

LOCTITE® SF 7649 Primers, Henkel Corporation

Supplier: Henkel
Loctite®
209715
76313-914CS 435.15 USD InStock
76313-914
LOCTITE® SF 7649 Primers, Henkel Corporation
Adhesives
Designed to promote the curing speed of LOCTITE® anaerobic adhesives and sealants, without any significant loss of joint strength.

  • Recommended for applications with passive metals or inert surfaces and with large bond gaps

LOCTITE® SF 7649 primers are particularly recommended when the application temperature is below 15 °C.

Ordering information: Ten pieces per case. Must be ordered in case quantity.

Packaging: 4.5 oz. Aerosol Can, 0.246 kg Package Weight, 5.6L×2.17W×2.17H cm Packaging Dimension.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty