Would you like to log in again?
Multifuge™ X4 Pro-MD, Thermo Scientific
Supplier: Thermo Scientific
75009500 75016046CAP 75016058 75009515 75016046 75009516CAP 75016057 75009501CAP 75016034 75016033CAP 75009615 75016067 75016033 75009501 75016045CAP 75016066 75009516 75016034CAP 75016045
76326-282EA 20576.73 USD InStock
76326-282 76326-394 76326-250 76326-242 76326-220 76326-230 76326-222 CA76501-600 76326-244 76326-254 CA76501-598 76326-234 76326-256 CA76501-564 76326-236 76326-228 CA76501-562 CA76501-592 CA76501-590
Multifuge™ X4 Pro-MD, Thermo Scientific
Centrifuges Benchtop Centrifuges
Process a range of general-purpose applications including cell culture, microplate, bioproduction, blood separation, and microbiology with the Thermo Scientific™ Multifuge™ X4 Pro Centrifuge series featuring our new touch screen technology, saves time and makes daily tasks easier.
  • Modern, intuitive touchscreen interface enables easy access and programming for faster results
  • Lighthouse mode: Easily monitor run progress from across the laboratory with enlarged time and tracker bar, end-of-run alerts can be programmed with customised volume settings
The Multifuge™ X4 Plus & Pro series can process up to 40×50ml conical tubes, 196×5/7 ml blood tubes, 24 micro plates.
The Megafuge™ X4 Plus & Pro general-purpose centrifuge series can process clinical protocols, cell culture applications, and microplate processing. The refrigerated version is designed for handling temperature-sensitive sample processing between –10 and 40 °C.
Auto-Lock™ III Rotor system allows easy push-button installation and exchange of rotors, as well as easy access to the rotor chamber for quick cleaning.
Thermo Scientific Fiberlite carbon fiber rotors offer an advanced alternative to metal centrifuge rotors, delivering superior performance without fatigue or corrosion for safe and worry-free operation spin after spin. They enhance productivity by reducing run times with higher G-forces; their corrosion- and fatigue-resistant design ensures structural integrity, and your investment is secured with a 15-year warranty.
ClickSeal™ Bucket Sealing System seals with a snap, eliminating screws and complicated clips. It is glove-friendly and certified for Biocontainment by CAMR in Porton Down, UK.
SMARTSpin™ Technology is an advanced rotor management system that maximizes acceleration, braking, and residual load imbalances for each rotor and bucket option. The system optimizes safety and improves separations.
Certifications: Certifications EN/UL/IEC 61010-1, EN/UL/IEC 61010-2-020 and 61010-2-101, EN/UL/IEC 61326-1, EN/UL/IEC 61326-2-6, FCC part 15.
Delivery information: All packages include: TX-1000 rotor cross, TX-1000 buckets (set of 4), ClickSeal biocontainment lids (set of 4)
Cell culture package includes 50 ml adapter (set of 4), 15 ml adapter (set of 4)
Blood tube package includes: 5/7ml adapter (set of 4), 10 ml adapter (set of 4)
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty