Would you like to log in again?
Bottles with Cap Assemblies, Beckman Coulter
Supplier: Beckman Coulter
355603 356011 357000 356013 356011 356013 357001 357000 355607 357001 355605 361691 355603
76335-396BX 147.16 USD InStock
CABK356013 76335-396 CABK356011 CABK357001 CABK357000 BK357000 BK356011 BK357001 BK-355603 BK356013 BK361691 BK355607 BK355605
Bottles with Cap Assemblies, Beckman Coulter
Bottles Centrifuge Bottles
Bottle assemblies are for use in high-performance and high-capacity rotors.
  • Transparent
  • Available in polycarbonate and polyallomer
  • Also works in benchtop centrifuges and microcentrifuges
Bottles with assemblies can also be used in benchtop centrifuges and microcentrifuges. Bottles are transparent and available in two material options.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty