Would you like to log in again?
TUBING ADVANTASIL HP 0.250 X 0.375 IN
Supplier: NEWAGE INDUSTRIES. INC
Tubing, Advantasil High Pressure Silicone, Increase flow rate, reduce processing time with APHP unreinforced for high pressure application, Flexible, translucent, durable, and ideal for Single-Use, Dimensions: 0.250 X 0.375 in
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
TUBING ADVANTASIL HP 0.250 X 0.375 IN 76407-462 A7040553-50 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty