Synonyms:
Dihydrogen oxide
W3340
W3345
W3340
W3350
W3330
W3330
W3335
W3335
W3345
W3350
100216-650EA
77.15
USD
InStock
CA100219-594
100216-650
100219-594
CA100216-650
76423-628
CA100219-942
100220-288
CA100220-288
CA100219-912
76423-626
Water, Molecular Biology Grade, PCR certified DNase/RNase tested, sterile
Water
Molecular Biology Grade Water is PCR certified negative for DNAse and RNAse (sensitivity to femtomoles) and is negative for non-specific Endonuclease, Exonuclease and Protease activity.