Really versatile general purpose apolar phase for a wide range of MS applications, including (for example): PCBs, simulated distillation, gases, essential oils, hydrocarbons analysis, pesticides, semi volatiles, oxygenates. HI-1 MS is an MS grade low-bleeding cross bonded column and assure a long lifetime even under high temperature limits.
- Phase composition: Cross bonded 100% methyl polysiloxane (100% dimethylpolysiloxane) - MS grade
- Long lifetime
- Equivalent to USP phases G1, G2, G38
Max temperature limits may change depending on the film thickness.