Very popular PEG high polarity MS phase for a wide range of applications, including (for example): essential oils, flavors and fragrances, alkenols. HI-WAX MS is an MS grade low-bleeding crossbonded column.
- Phase composition: Crossbonded Polyethylene Glycol (PEG) - MS grade
- Long lifetime
Maximum temperature limits may change depending on the film thickness.