Would you like to log in again?
Socorex® Qualitix® Premium MicroPipette Tips, DWK Life Sciences
Supplier: DWK Life Sciences
Qualitix®
3070.0200PR 3090.0010PR 3070.1000PRS 3070.1000R 308.0350B 3080.0200GR 3070.0200R 3070.0200FRS 302.0010GR 3070.1000T 3070.1000FRS 3070.1000B 3070.0200PRS 3080.0020FRS 3090.0010B 3090.0010FRS 3070.1000PR 3090.0010T 3070.0200B 3090.0010PRS 308.0350R 3070.0200T 3090.0010R 3080.0100FRS 308.0300FR
76437-372CS 135.6 USD InStock
76437-372 76437-350 76437-370 76437-354 76437-374 76437-330 76437-352 76437-348 76437-326 76437-368 76437-346 76437-324 76437-328 76437-340 76437-360 76437-322 76437-366 76437-344 76437-364 76437-342 76437-358 76437-336 76437-356 76437-334 76437-338
Socorex® Qualitix® Premium MicroPipette Tips, DWK Life Sciences
Pipette Tips
The Qualitix® Pipette tip program is all about quality, flexibility and compatibility.
  • Wide selection, superior quality, autoclavable
  • Bags, racks and tipfill rack filling system
  • Lot by lot consistency
  • RNase-, DNase- and pyrogen-free
  • Reduced packaging size, less bench space requirement
  • Broad compatibility with major micropipette brands
The NEW Qualitix® Pipette Tips offer accurate liquid delivery in both standard and low protein binding resin, fitting not only Socorex® but all major micropipette brands. Packaging selection (bags, racks and tipfill rack filling system), together with a choice of tip styles with and without protection filter offer appreciated versatility. Qualitix® and Socorex® micropipettes are the most perfect match for optimized test results according to international standards.
Certifications: Certified free of nuclease, endotoxin and ATP.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty