High Performance FR Lightweight Pants feature 8 oz. Westex® UltraSoft® fabric that has a unique basketweave blend and provides increased mobility and comfort, making them an excellent choice for arc flash and flash fire protection.
- Wider waistband for increased comfort
- Articulated knees offer better mobility
- Cell phone pocket on right leg
- Stealth zipper pocket on right leg
- Crotch gusset for increased comfort
- Extra material in heal to reduce wear
Lakeland FR Lightweight Pants are made with Westex® UltraSoft® and are NFPA 2112, UL certified, and meet NFPA 70E, CAT 2, making them an excellent choice for arc flash and flash fire protection. The unique basketweave fabric blend provides increased mobility and comfort. Attention to design with features like an articulated knee, wider waistband, and a crotch gusset ensure all day comfort.
Certifications: NFPA 70E. NFPA 2112 - UL certified. ATPV: 9.8 cal/cm². CAT: 2.