Would you like to log in again?
BHD Autoclave Bags with Self-Seal Closure, Blue, Keystone Cleanroom Products
Supplier: Keystone Cleanroom Products
BNW1648SS2P BNW1622SS2P BNW2226SS2P BNW1240SS2P BNW2626SS2P BNW0638SS2P BNW2832SS2P BNW0844SS2P BNW4448SS2P BNW2646SS2P BNW0840SS2P BNW4446SS2P BNW4040SS2P BNW5858SS2P
76451-528CS 2012.31 USD InStock
76451-528 76451-538 76451-526 76451-548 76451-536 76451-546 76451-534 76451-544 76451-532 76451-542 76451-530 76451-552 76451-540 76451-550
BHD Autoclave Bags with Self-Seal Closure, Blue, Keystone Cleanroom Products
Bags Sterilization Pouches
Self-sealing BHD Autoclave bags can be used as either the inner, or outer bag for the steam sterilization of pharmaceutical change parts.
  • Manufactured and packaged in a controlled cleanroom environment
  • Reduces labor costs
  • Simplifies the conformance to wrapping SOP's
  • Autoclavable up to 135 °C
  • Tear and puncture resistant
  • Significant process improvement compared to wrapping with BioShield paper
  • Eliminates need to cut material and the use of autoclave tape
Standardize and simplify the way equipment is wrapped for steam sterilization.
The use of these 2 ply self-sealing bags will also save valuable time in the equipment prep area. The 2-ply BHD material resists ripping and punctures.
Since 100% of the surface area allows for steam penetration and drying, each load can be configured for maximum efficieny and throughput.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty