Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Equipment GMP Covers, 1422A Tyvek®, Autoclavable, Keystone Cleanroom Products

Supplier: Keystone Cleanroom Products
502872Z2TWWP4 COVTVK642460 COVTVK603072E COVTVK362660E COVTVK742686E COVTVK443632 COVTVK603072 COVTVK505036E COVTVK642478E COVTVK482860E COVTVK381402E COVTVK444460 COVTVK602472 COVTVK302212E COVTVK302456Z1 COVTVK482860 COVTVK642460E COVTVK545486 COVTVK545486E COVTVK250304E COVTVK222236E COVTVK505050E 663068TWEWP COVTVK382862Z2 COVTVK1024034E COVTVK762672E COVTVK141008 COVTVK262010E COVTVK505050 COVTVK602472E COVTVK342604E COVTVK281006E COVTVK423460 COVTVK423460E 462660Z2WT4WP COVTVK210304E COVTVK200805E COVTVK604444 COVTVK342233 COVTVK1228688 COVTVK322824 COVTVK604444E COVTVK642478 COVTVK682238E COVTVK602854E COVTVK642660Z2 COVTVK251603E 663852TWEWP COVTVK605060 COVTVK563660E COVTVK643068E COVTVK1024034 COVTVK605060E COVTVK502474E COV-TVK-201810E
76451-704CS 7342.31 USD InStock
76451-704 76451-702 76451-668 76451-700 76451-666 76451-664 76451-662 76451-660 76451-728 76451-736 76451-658 76451-734 76451-656 76451-732 76451-698 76451-654 76451-730 76451-696 76451-652 76451-694 76451-650 76451-692 76451-690 76451-718 76451-716 76451-726 76451-648 76451-724 76451-646 76451-722 76451-688 76451-644 76451-720 76451-686 76451-642 76451-684 76451-640 76451-682 76451-680 76451-708 76451-706 76451-638 76451-714 76451-636 76451-712 76451-678 76451-634 76451-710 76451-632 76451-676 76451-674 76451-630 76451-672 76451-670 76918-240
Equipment GMP Covers, 1422A Tyvek®, Autoclavable, Keystone Cleanroom Products
Covers
Save time and standardize the activities in equipment preparation for steam sterilization; or use in any pharmaceutical manufacturing setting to keep cleaned equipment protected from contamination.

  • Manufactured and packaged in a controlled cleanroom environment
  • Reduces labor costs
  • Simplifies the conformance to wrapping SOP's
  • Individually poly bagged and labeled
  • Steam sterilization indicator - autoclavable at 121 to 125 °C
  • Tear and puncture resistant
  • Hydrophobic
  • Lint-free

These covers provide a convenient, time-saving method of wrapping pharmaceutical production equipment prior to steam sterilization. These covers allow immediate steam penetration and work best with modern pulse vacuum autoclaves for sufficient drying.

Their use as cleaned equipment coverings,in all pharmaceutical manufacturing environments, helps minimize microbial and particulate contamination risks.

Certifications: Certificates of conformance sent with each shipment.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty