VWR® Made in USA SMS Coveralls are an ideal option for single-use needs.
- 3-layer SMS construction
- Serged seams
- Produced with latex-free elastic
- Made in USA
Coveralls are manufactured from a unique multi-layered spunbonded-meltblown-spunbonded (SMS) fabric that offers high tensile strength while remaining soft, comfortable, and breathable.
They provide light chemical splash resistance. The material is low-linting and resistant to tears and punctures. Ideally suited for use in high-class protection areas of medical, industrial, laboratory, electronic, and pharmaceutical environments.
Ordering information: Also available in the Made in USA PPE line are masks, lab coats, scrubs, and SMS coveralls.
Packaging: Packed 25 coveralls per case.