The VWR® Made in USA PPE line has been expanded to include microporous material coveralls.
- Microporous, 60 gsm, two-layer material
- Fluid and particulate barrier
- Excellent breathability
- Serged seams
- Hood and Boot options
- Elastic or open cuff and ankle options
Microporous material coveralls provide increased fluid protection with great comfort. Choose from a variety of styles, including different wrist and ankle options, with or without hoods, boots, or both.
Ordering information: Also available in the Made in USA PPE line are masks, lab coats, scrubs, and SMS coveralls.
Packaging: Packed 25 coveralls per case.