Would you like to log in again?
Cell Meter™ Fluorimetric Mitochondrial Superoxide Activity Assay Kit*Optimized for Flow Cytometry*, AAT Bioquest
Supplier: AAT Bioquest
22970
76483-834EA 361.89 USD InStock
76483-834
Cell Meter™ Fluorimetric Mitochondrial Superoxide Activity Assay Kit*Optimized for Flow Cytometry*, AAT Bioquest
Assays Cellular Assays
Mitochondria are major producers of cellular superoxide.
  • Ready to use
  • Contains all necessary components
  • Robust and adaptable
  • Optimized for flow cytometry but suitable for all major fluorescence platforms
  • Results with only 1 hour incubation
Caution: For research use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty