Would you like to log in again?
Spexyte™ Intracellular pH Calibration Buffer Kit, AAT Bioquest
Supplier: AAT Bioquest
21235
76484-784EA 240.68 USD InStock
76484-784
Spexyte™ Intracellular pH Calibration Buffer Kit, AAT Bioquest
Assays Cellular Assays
Intracellular pH (pHi) plays an important modulating role in many cellular events, including cell volume regulation, cellular metabolism, calcium regulation, receptor-mediated signal transduction, ion transport, endocytosis, and other cellular processes.
  • Suitable for all major pH indicators
  • Sensitive and quick results
  • Includes a range of calibration buffers
Caution: For research use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty