Would you like to log in again?
Ultra Low Temperature Freezers, –86 °C, Thermo Scientific
Supplier: Thermo Scientific
TDE50086FD TDE50086FA5ID TDE30086LD TDE30086FD TDE30086FD5ID TDE40086LA TDE40086FD TDE60086LA TDE40086FD5ID TDE30086LA TDE60086FA TDE40086FA5ID TDE50086LD TDE60086FA5ID TDE50086FA TDE50086LA TDE30086FA TDE60086LD TDE60086FD TDE40086LD TDE30086FA5ID TDE40086FA TDE60086FD5ID TDE50086FD5ID
76535-070EA 31467.26 USD InStock
76535-070 76595-480 76535-060 76535-050 76535-072 76535-062 76595-484 76595-574 76595-532 76535-056 76595-564 76535-068 76535-046 76535-058 76595-622 76535-048 76535-052 76595-524 76595-612 76535-042 76535-064 76535-054 76535-044 76535-066
Ultra Low Temperature Freezers, –86 °C, Thermo Scientific
Freezers Ultra Low Temperature Freezers
Dependable –86 °C storage, powered by H-drive for operational savings and sustainability.
  • Low energy
  • Low peak variation
  • Fast recovery times after door opening
  • Long warm up time
  • Water blown VIP insulation
  • USB data download capability
  • Choice of 4 sizes
Designed to keep your samples safe. Limit environmental impact as units have Green hydrocarbon refrigerants, are user friendly and quiet.
Certifications: All models are SNAP Compliant with ACT Label. FA/FD are general purpose models, LA/LD are FDA Class II models.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty