VWR® lab benches are ergonomically designed to meet the needs of laboratory personnel.
- Snow white frame
- Legged steel structure-4
- Load capacity 2000 lbs. for even the toughest environments
- Can be easily equipped with accessories
- ISO class 7 cleanroom compatible
There are many possible adjustments, all components fit together seamlessly, and each detail is designed for real world functionality. The lab bench adapts to the work process and the needs of the user. As a result, all movements of the user can be optimized, which in turn increases performance and productivity and decreases work-related stress. All bench frames can be equipped with casters to create a mobile workstation solution.
Our 4-leg frame height range can be adjusted from 27.56 - 40.55", in 1" increments, making these tables suitable for work while sitting or standing. Shelves depth 11.81".
Ordering information: Notched work surfaces add 2.625" to depth of table frame. Work surfaces for single bay uprights include two notches (left and right); work surfaces for double bay uprights include three notches (left, center, right).
Tables ship LTL, flat-packed. Installation not included.
Packaging: Benches are shipped knocked down, on non-stackable pallets. Assembly instructions and hardware included.