Would you like to log in again?
Sol-Guard® Safety TB Syringes, Sol M
Supplier: Sol-Millennium
200034SG 2131516SG 200018SG 2031516SG 200067SG 200088SG
76582-670BX 38.55 USD InStock
76582-670 76582-672 76582-678 76582-668 76582-710 76582-666
Sol-Guard® Safety TB Syringes, Sol M
Syringes Syringes, Medical
Sol-Guard safety syringes have a fixed needle and safety shield attached, to reduce the risk of needlestick injuries.
  • Sliding Safety Shield with easy grip ridges to protect against needlestick injuries
  • Transparent shield for easy sighting of the needle tip
  • Transport position – eliminate the need to recap the needle after drawing-up medication by sliding up the safety shield
  • Clear and bold scale markings
Low dead space design reduces waste and allows for more efficient medication delivery. A transparent safety shield slides up to cover the needle during transport and after medication delivery, reducing exposure to the needle throughout the entire injection process.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty