Would you like to log in again?
TDE Series Ultra-Low Temperature Freezers with LN₂ Back-Up System, Thermo Scientific
Supplier: Thermo Scientific
TDE40086LDRLN TDE50086LARLN TDE60086FDRLN TDE50086FDRLN TDE40086FARLN TDE50086LDRLN TDE60086LARLN TDE60086LDRLN TDE40086LARLN TDE50086FARLN TDE40086FDRLN TDE60086FARLN
76595-590EA 27341.15 USD InStock
76595-590 76595-638 76595-550 76595-572 76595-582 76595-530 76595-598 76595-620 76595-630 76595-544 76595-646 76595-538
TDE Series Ultra-Low Temperature Freezers with LN₂ Back-Up System, Thermo Scientific
Freezers Ultra Low Temperature Freezers
TDE series ultra-low temperature freezers are part of the Standard Performance (STP) family.
Insulation: water-blown polyurethane foam
Certifications: cULus; FDA Class II.
Delivery information: Package includes one ULT freezer, two shelves of sliding drawer racks and boxes and a factory-installed LN2 back-up system.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty