Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

TDE Series Ultra-Low Temperature Freezers, −40 °C, Thermo Scientific

Supplier: Thermo Scientific
TDE50040LD TDE60040FD TDE60040LD TDE50040FD TDE50040LA TDE40040LD TDE60040FA TDE50040FA TDE40040LA TDE40040FA TDE60040LA TDE40040FD
76595-606EA 16485.29 USD InStock
76595-606 76595-608 76595-508 76595-492 76595-560 76595-552 76595-562 76595-600 76595-610 76595-500 76595-558 76595-516
TDE Series Ultra-Low Temperature Freezers, −40 °C, Thermo Scientific
Freezers Ultra Low Temperature Freezers
TDE series −40 °C ultra-low temperature freezers, powered by H-drive, features three upright models, maximizing storage capacity from 40000 up to 60000 2 ml vials.

  • H-drive hydrocarbon refrigeration system
  • Low peak variation
  • User-friendly and quiet (sound output below 53 dBA)
  • Security features includes alarm settings and padlock-compatible doors
  • Limit environmental impact and reduces HVAC costs

The TDE series is a sustainable and reliable solution with ultimate sample security and operational savings for every lab. All models contains four inner doors allowing for easy sample access without exposing greater freezer content to ambient conditions

Improved customer convenience with new ergonomically designed handle, high contrast eye-level display and comfortable easy-access shelf locations. Compatible with most racking systems including a full range of Thermo Scientific™ racks for boxes, microplates, Thermo Scientific™ Matrix™ and Nunc CryoBank™ tubes.

Manufactured in an award-winning, zero-waste to landfill facility. The natural refrigerants and water-blown foam insulation, making the series compliant with the U.S. Environmental Protection Agency’s Significant New Alternatives Policy (SNAP), the European Union’s F-Gas and other sustainability standards.

Certifications: CE and cULus, FDA Class II.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty