Would you like to log in again?
Anti-Smooth muscle myosin Mouse Monoclonal Antibody [clone: SMMS-1]
Supplier: Diagnostic BioSystems
MOB467-05 MOB467 MOB467-01 MOB467-01-RUO MOB467-05-RUO MOB467-RUO
76633-980EA 581.62 USD InStock
CA76633-980 76633-980 CA76633-978 76633-978 CA76633-976 76633-976
Anti-Smooth muscle myosin Mouse Monoclonal Antibody [clone: SMMS-1]
Antibodies
Monoclonal anti-smooth muscle myosin reacts with myosin heavy chain polypeptides of 200 and 204 kDa. It does not react with skeletal, cardiac, or non-muscle myosin. In immunohistochemistry it stains vascular and visceral smooth muscle cells and cells that have smooth muscle-like characteristics.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty