Would you like to log in again?
Anti-Actin Muscle Mouse Monoclonal Antibody [clone: HHF35]
Supplier: Diagnostic BioSystems
PDM002-RUO PDM002
76634-256EA 269.92 USD InStock
76634-256 CA76634-256
Anti-Actin Muscle Mouse Monoclonal Antibody [clone: HHF35]
Antibodies
This pan actin is useful in identifying tumors arising from muscle cells, i.e., leiomyosarcoma as well as rhabdomyosarcoma.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty