Would you like to log in again?
Anti-PSAP Mouse Monoclonal Antibody [clone: PASE/4LJ]
Supplier: Diagnostic BioSystems
PDM037-RUO PDM037
76634-294EA 284.24 USD InStock
76634-294 CA76634-294
Anti-PSAP Mouse Monoclonal Antibody [clone: PASE/4LJ]
Antibodies
This antibody is specific to human prostatic acid phosphatase. It reacts with the glandular epithelium of normal and hyperplastic prostate and with prostate adenocarcinoma.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty