Would you like to log in again?
Anti-CD68 Macrophage Mouse Monoclonal Antibody [clone: KP1]
Supplier: Diagnostic BioSystems
PDM066 PDM066-RUO
76634-322EA 269.92 USD InStock
76634-322 CA76634-322
Anti-CD68 Macrophage Mouse Monoclonal Antibody [clone: KP1]
Antibodies
This antibody is specific to macrophages in a wide variety of human tissues. It reacts with myeloid precursors and peripheral blood granulocytes. It also stains a cell population known as plasmacytoid T cells.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty