Would you like to log in again?
Anti-Mart-1/Melan A Mouse Monoclonal Antibody [clone: A103]
Supplier: Diagnostic BioSystems
PDM153 PDM153-RUO
76634-414EA 391.73 USD InStock
CA76634-414 76634-414
Anti-Mart-1/Melan A Mouse Monoclonal Antibody [clone: A103]
Antibodies
This antibody is specific to a protein of 20 kDa known as MART-1 (melanoma antigen recognized by T cells-1 or Melan-A. This antibody does not cross-react with MAGE-1 or tyrosinase. MART-1 is a newly identified melanocyte differentiation antigen recognized by autologous cytotoxic T lymphocytes. This antibody stains melanomas and other tumors showing melanocytic differentiation.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty