Would you like to log in again?
Anti-Mast Cell Tryptase Mouse Monoclonal Antibody [clone: CC1]
Supplier: Diagnostic BioSystems
PDM159-RUO PDM159
76634-422EA 379.79 USD InStock
76634-422 CA76634-422
Anti-Mast Cell Tryptase Mouse Monoclonal Antibody [clone: CC1]
Antibodies
This antibody reacts with mast cells distributed in skin, synovium, lung, and heart. Mast cells contain a number of preformed chemicals mediators such as histamine, chymase, carboxypeptidase, and proteolytic tryptase. Human mast cell chymase is considered to be an important marker of mast cells and is an important mediator of inflammation.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty