This model is a one-piece molded cassette so no separate lids to manage. It has a front opening and a 45° smooth writing surface. It can be opened and closed up to five times guaranteed, securely relocking to avoid specimen loss.
- Unique lateral vents, fluids can access tissue from all four sides
- Basket style for small tissue management
- One-piece molded with front opening
- Smooth 45° angle writing surface
- Available in 11 colors
Molded from high-density polymer, it keeps specimens safely submerged in liquid.
Available for tissue, biopsy and micro-biopsy. This cassette model is supplied in bulk and also in taped stacks for use with Leica and Sakura Ink Jet printers.
Certifications: IVDR, CE.