Would you like to log in again?
Human/Mouse/Rat/Cynomolgus/Rhesus Recombinant Activin A/INHBA Protein (from HEK293 Cells)
Supplier: Sino Biological
10429-HNAH-100 10429-HNAH-1 10429-HNAH-20
76777-812EA 498 USD InStock
76777-812 76778-042 76641-830
Human/Mouse/Rat/Cynomolgus/Rhesus Recombinant Activin A/INHBA Protein (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding mature form of human / mouse / rat / cynomolgus / rhesus INHBA (NP_002183.1) (Gly311-Ser426) was expressed. The mature form sequences of the human, mouse, rat, cynomolgus and rhesus INHBA are identical.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty