Would you like to log in again?
TET-dT Phosphoramidite
Supplier: AAT BIOQUEST INC
6212 6213
76644-688EA 194.16 USD InStock
76644-688 76644-690
TET-dT Phosphoramidite
TET-dT Phosphoramidite
TET-dT phosphoramidite contains a DMT group to allow quantification of coupling. It can be inserted into a target sequence as a replacement for a dT residue. It provides a valuable tool to add the TET tag to a desired location of an oligo sequence. It is complimentary to the commonly used 6-TET phosphoramidite that contains no DMT group and can only be added at the 5'-terminus, thus terminating synthesis. AAT Bioquest also offer fluorescein-dT phosphoramidite, HEX-dT phosphoramidite, biotin-dT phosphoramidite, biotin-dT CPG and Fluorescein-dT CPG.
  • For efficiently labeling an oligonucleotide
Caution: Research Use Only (RUO)
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty