Would you like to log in again?
Oligo Pro II System Capillary Arrays
Supplier: Agilent
M5340AA-001 A2500-1275-5580 A2500-9675-5580 M5340AA-001 A2500-2475-5580 A2500-9675-5580 M5340AA-003 M5340AA-002 M5340AA-002 A2500-2475-5580 A2500-1275-5580 M5340AA-003
76645-486EA 2928.3 USD InStock
76645-486 76645-188 76645-274 76645-130 76645-042 CA76645-280 76645-280 CA76645-130 CA76645-042 CA76645-274 CA76645-188 CA76645-486
Oligo Pro II System Capillary Arrays
Electrophoresis Systems Capillary Electrophoresis Systems
Three different capillary arrays for analysis on the Oligo Pro II system.
  • High resolution - optimized length allowing for n-1 resolution through 60 nt
  • Variable throughput - 12, 24, and 96 capillary arrays designed to meet individual throughput demands
  • Designed for the Oligo Pro II system - compatible with UV detection allowing for visualization of unlabeled oligonucleotides
The Oligo Pro II system has 3 capillary arrays, the 12-, 24-, and 96-Capillary Array Long, for analysis with the Oligo Pro Reagents. All three have a separation or effective length of 55 cm, with a total length of 80 cm. The 12 parallel capillary array is ideal for lower throughput, while the 24 and 96 are designed for medium to high-throughput laboratories.
Caution: For research use only. Not for use in diagnostic procedures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty