Would you like to log in again?
Femto Pulse System
Supplier: Agilent
M5330AA FP-OQ M5330AA FP-OQ
76645-432EA 1161 USD InStock
CA76645-432 76645-432 76645-176 CA76645-176
Femto Pulse System
Electrophoresis Systems Capillary Electrophoresis Systems
Pulsed-field capillary electrophoresis system for QC of high molecular weight DNA and RNA samples.
  • Large fragment resolution - separate high molecular weight DNA smears and fragments through 165 kb in about 1.5 hours
  • Femtogram level sensitivity - achieve 10 times higher sensitivity for nucleic acid smears and up to 100 times higher for nucleic acid fragments
  • Simple sample preparation - minimize sample degradation with fewer pipetting steps and stable reagents
  • Powerful analysis software - one intuitive software program automates analysis of diverse sample types
  • Conserve precious sample - unparalleled sensitivity allows you to use less precious sample and achieve superior detection
  • Make workflows more efficient - eliminate pulsed-field gel electrophoresis from long-read NGS library preparation and BAC analysis
  • Desktop PC supplied with a comprehensive 3-year warranty through HP
By using a pulsed-field power supply, large molecular weight DNA fragments can be separated and nucleic acids detected into the femtogram range. Compared to other non-denaturing gel-based analytical instruments, the Femto Pulse system easily achieves 10 times higher sensitivity for nucleic acid smears and up to 100 times higher sensitivity for nucleic acid fragments.
The ability to separate high molecular weight DNA up to 165 kb and detect nucleic acids down to 50 fg/µl input concentration make the Femto Pulse system ideal for long-read NGS QC, gDNA, small RNA, or cfDNA analysis from low concentration samples.
Caution: For research use only. Not for use in diagnostic procedures.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty